siRNA Details
| Gene Symbol: Sugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108397.1 | NP_001101867.1 | 3 | 1901 | ||
| XM_008771124.1 | XP_008769346.1 | 2 | 1488 | |||
| XM_008771125.1 | XP_008769347.1 | 3 | 1528 | |||
| XM_008771126.1 | XP_008769348.1 | 3 | 1663 | |||
| XM_008771127.1 | XP_008769349.1 | 2 | 1488 | |||
| XM_008771128.1 | XP_008769350.1 | 3 | 1563 | |||

Sugp2
Genomic Map