siRNA Details
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135756.1 | NP_001129228.1 | 9 | 1428 | ||
XM_008771227.1 | XP_008769449.1 | 8 | 2082 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135756.1 | NP_001129228.1 | 9 | 1428 | ||
XM_008771227.1 | XP_008769449.1 | 8 | 2082 |