siRNA Details
| Gene Symbol: Primpol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253167.2 | XP_006253229.1 | 5 | 471 | ||
| XM_341433.6 | XP_341434.3 | 5 | 468 | |||
| GenBank mRNA | FQ230438.1 | - | 2 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Primpol| Gene Symbol: Primpol | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253167.2 | XP_006253229.1 | 5 | 471 | ||
| XM_341433.6 | XP_341434.3 | 5 | 468 | |||
| GenBank mRNA | FQ230438.1 | - | 2 | 94 | ||