siRNA Details
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253167.2 | XP_006253229.1 | 6 | 687 | ||
XM_341433.6 | XP_341434.3 | 6 | 684 | |||
GenBank mRNA | FQ230438.1 | - | 3 | 310 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Primpol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253167.2 | XP_006253229.1 | 6 | 687 | ||
XM_341433.6 | XP_341434.3 | 6 | 684 | |||
GenBank mRNA | FQ230438.1 | - | 3 | 310 |