siRNA Details
Gene Symbol: Eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014143.1 | NP_001014165.1 | 3 | 582 | ||
GenBank mRNA | BC089828.1 | - | 3 | 582 | 7322740 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014143.1 | NP_001014165.1 | 3 | 582 | ||
GenBank mRNA | BC089828.1 | - | 3 | 582 | 7322740 |