siRNA Details
| Gene Symbol: Eri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014143.1 | NP_001014165.1 | 6 | 859 | ||
| GenBank mRNA | BC089828.1 | - | 6 | 859 | 7322740 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eri1| Gene Symbol: Eri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014143.1 | NP_001014165.1 | 6 | 859 | ||
| GenBank mRNA | BC089828.1 | - | 6 | 859 | 7322740 | |