siRNA Details
| Gene Symbol: Alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108401.1 | NP_001101871.1 | 3 | 870 | ||
| XM_006253354.2 | XP_006253416.1 | 3 | 1142 | |||
| XM_006253355.2 | XP_006253417.1 | 3 | 844 | |||
| XM_006253356.2 | XP_006253418.1 | 3 | 1467 | |||

Alg11
Genomic Map