siRNA Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127301.1 | NP_001120773.1 | 6 | 602 | ||
| XM_008771401.1 | XP_008769623.1 | 5 | 1490 | |||
| GenBank mRNA | BC166799.1 | - | 6 | 602 | 7938235 | |
| FQ210009.1 | - | 6 | 601 | |||

Cul4a
Genomic Map