siRNA Details
| Gene Symbol: Ankrd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253453.2 | XP_006253515.1 | 6 | 1215 | ||
| XM_006253454.2 | XP_006253516.1 | 6 | 1556 | |||
| XM_006253458.2 | XP_006253520.1 | 6 | 1655 | |||
| XM_006253459.2 | XP_006253521.1 | 4 | 1355 | |||
| XM_008771402.1 | XP_008769624.1 | 6 | 5363 | |||
| GenBank mRNA | BC098779.1 | - | 2 | 78 | ||

Ankrd10
Genomic Map