siRNA Details
| Gene Symbol: Usp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001516.2 | NP_001001516.1 | 20 | 2983 | ||
| XM_006243793.2 | XP_006243855.1 | 21 | 3301 | |||
| XM_006243794.2 | XP_006243856.1 | 21 | 3298 | |||
| XM_006243795.2 | XP_006243857.1 | 21 | 3295 | |||
| XM_006243796.2 | XP_006243858.1 | 21 | 3277 | |||
| XM_006243797.2 | XP_006243859.1 | 21 | 3300 | |||
| XM_006243798.2 | XP_006243860.1 | 21 | 3297 | |||
| XM_006243799.2 | XP_006243861.1 | 21 | 3294 | |||
| GenBank mRNA | AY605065.1 | - | 20 | 2983 | ||
| BC168243.1 | - | 20 | 3309 | |||

Usp19
Genomic Map