siRNA Details
| Gene Symbol: Nsun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108403.1 | NP_001101873.1 | 6 | 646 | ||
| XM_006227792.2 | XP_006227854.1 | 5 | 581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsun2| Gene Symbol: Nsun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108403.1 | NP_001101873.1 | 6 | 646 | ||
| XM_006227792.2 | XP_006227854.1 | 5 | 581 | |||