siRNA Details
| Gene Symbol: Ercc6l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253498.2 | XP_006253560.2 | 19 | 3818 | ||
| XM_008771452.1 | XP_008769674.1 | 19 | 3400 | |||
| GenBank mRNA | FQ211532.1 | - | 19 | 3765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ercc6l2| Gene Symbol: Ercc6l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253498.2 | XP_006253560.2 | 19 | 3818 | ||
| XM_008771452.1 | XP_008769674.1 | 19 | 3400 | |||
| GenBank mRNA | FQ211532.1 | - | 19 | 3765 | ||