siRNA Details
| Gene Symbol: RGD1311345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173436.1 | NP_001166907.1 | 4 | 953 | ||
| GenBank mRNA | BC098862.1 | - | 2 | 155 | ||
| BC168185.1 | - | 4 | 667 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311345| Gene Symbol: RGD1311345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173436.1 | NP_001166907.1 | 4 | 953 | ||
| GenBank mRNA | BC098862.1 | - | 2 | 155 | ||
| BC168185.1 | - | 4 | 667 | |||