siRNA Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014144.1 | NP_001014166.1 | 2 | 302 | ||
| XM_006253833.2 | XP_006253895.1 | 3 | 428 | |||
| GenBank mRNA | AY325171.1 | - | 316 | |||
| BC088287.1 | - | 2 | 302 | 7321485 | ||

Adtrp
Genomic Map