siRNA Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014144.1 | NP_001014166.1 | 1 | 180 | ||
| XM_006253833.2 | XP_006253895.1 | 2 | 306 | |||
| GenBank mRNA | AY325171.1 | - | 107 | |||
| BC088287.1 | - | 1 | 180 | 7321485 | ||

Adtrp
Genomic Map