siRNA Details
| Gene Symbol: Serpinb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007732.1 | NP_001007733.1 | 5 | 613 | ||
| XM_006253872.2 | XP_006253934.1 | 5 | 582 | |||
| GenBank mRNA | BC079063.1 | - | 5 | 613 | 7121006 | |
| FQ226618.1 | - | 2 | 148 | |||

Serpinb9
Genomic Map