siRNA Details
| Gene Symbol: Serpinb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007732.1 | NP_001007733.1 | 3 | 384 | ||
| XM_006253872.2 | XP_006253934.1 | 3 | 353 | |||
| GenBank mRNA | BC079063.1 | - | 3 | 384 | 7121006 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb9| Gene Symbol: Serpinb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007732.1 | NP_001007733.1 | 3 | 384 | ||
| XM_006253872.2 | XP_006253934.1 | 3 | 353 | |||
| GenBank mRNA | BC079063.1 | - | 3 | 384 | 7121006 | |