siRNA Details
| Gene Symbol: RGD1307443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197023.1 | NP_001183952.1 | 14 | 2458 | ||
| XM_006253936.2 | XP_006253998.1 | 14 | 3441 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307443| Gene Symbol: RGD1307443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197023.1 | NP_001183952.1 | 14 | 2458 | ||
| XM_006253936.2 | XP_006253998.1 | 14 | 3441 | |||