siRNA Details
| Gene Symbol: Cul2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108417.1 | NP_001101887.1 | 4 | 493 | ||
| XM_006254072.2 | XP_006254134.1 | 4 | 488 | |||
| XM_006254073.2 | XP_006254135.1 | 4 | 493 | |||
| XM_006254074.2 | XP_006254136.1 | 4 | 409 | |||

Cul2
Genomic Map