siRNA Details
| Gene Symbol: Fam208b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254220.2 | XP_006254282.1 | 6 | 703 | ||
| XM_006254221.2 | XP_006254283.1 | 6 | 884 | |||
| XM_006254222.2 | XP_006254284.1 | 6 | 445 | |||
| XM_006254223.2 | XP_006254285.1 | 6 | 703 | |||
| XM_006254224.2 | XP_006254286.1 | 6 | 448 | |||
| XM_341552.6 | XP_341553.3 | 6 | 704 | |||
| GenBank mRNA | BC168208.1 | - | 6 | 453 | ||

Fam208b
Genomic Map