siRNA Details
| Gene Symbol: Fam208b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006254220.2 | XP_006254282.1 | 10 | 1130 | ||
| XM_006254221.2 | XP_006254283.1 | 10 | 1311 | |||
| XM_006254222.2 | XP_006254284.1 | 10 | 872 | |||
| XM_006254223.2 | XP_006254285.1 | 10 | 1130 | |||
| XM_006254224.2 | XP_006254286.1 | 10 | 875 | |||
| XM_341552.6 | XP_341553.3 | 10 | 1131 | |||
| GenBank mRNA | BC168208.1 | - | 10 | 880 | ||

Fam208b
Genomic Map