siRNA Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170343.1 | NP_001163814.1 | 9 | 863 | ||
| XM_006254257.2 | XP_006254319.1 | 7 | 699 | |||
| GenBank mRNA | CD373126.1 | - | 5 | 544 | ||
| CO567453.1 | - | 39 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec61a2| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170343.1 | NP_001163814.1 | 9 | 863 | ||
| XM_006254257.2 | XP_006254319.1 | 7 | 699 | |||
| GenBank mRNA | CD373126.1 | - | 5 | 544 | ||
| CO567453.1 | - | 39 | ||||