siRNA Details
| Gene Symbol: Msrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031660.1 | NP_001026830.1 | 4 | 352 | ||
| GenBank mRNA | BC098953.1 | - | 4 | 352 | 7374871 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msrb2| Gene Symbol: Msrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031660.1 | NP_001026830.1 | 4 | 352 | ||
| GenBank mRNA | BC098953.1 | - | 4 | 352 | 7374871 | |