siRNA Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014147.1 | NP_001014169.2 | 4 | 358 | ||
| GenBank mRNA | BC079332.1 | - | 4 | 358 | 7096678 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpgs2| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014147.1 | NP_001014169.2 | 4 | 358 | ||
| GenBank mRNA | BC079332.1 | - | 4 | 358 | 7096678 | |