siRNA Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108434.1 | NP_001101904.1 | 3 | 391 | ||
| XM_006254850.2 | XP_006254912.1 | 3 | 2645 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fech| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108434.1 | NP_001101904.1 | 3 | 391 | ||
| XM_006254850.2 | XP_006254912.1 | 3 | 2645 | |||