siRNA Details
| Gene Symbol: Mppe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108435.2 | NP_001101905.1 | 4 | 778 | ||
| XM_006254879.2 | XP_006254941.1 | 4 | 772 | |||
| XM_006254881.2 | XP_006254943.1 | 4 | 1424 | |||
| GenBank mRNA | BC162043.1 | - | 4 | 778 | 7105580 | |

Mppe1
Genomic Map