siRNA Details
| Gene Symbol: LOC361346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017462.1 | NP_001017462.1 | 6, 7 | 1259 | ||
| XM_006254929.2 | XP_006254991.1 | 4, 5 | 1089 | |||
| XM_008772154.1 | XP_008770376.1 | 4, 5 | 1332 | |||
| GenBank mRNA | BC082051.1 | - | 6, 7 | 1259 | 7132090 | |

LOC361346
Genomic Map