siRNA Details
| Gene Symbol: LOC361346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017462.1 | NP_001017462.1 | 3 | 655 | ||
| XM_008772154.1 | XP_008770376.1 | 1 | 266 | |||
| GenBank mRNA | BC082051.1 | - | 3 | 655 | 7132090 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC361346| Gene Symbol: LOC361346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017462.1 | NP_001017462.1 | 3 | 655 | ||
| XM_008772154.1 | XP_008770376.1 | 1 | 266 | |||
| GenBank mRNA | BC082051.1 | - | 3 | 655 | 7132090 | |