siRNA Details
| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108440.1 | NP_001101910.1 | 10 | 1046 | ||
| XM_006255267.2 | XP_006255329.1 | 11 | 1581 | |||
| XM_006255268.2 | XP_006255330.1 | 11 | 1581 | |||
| XM_008772409.1 | XP_008770631.1 | 8 | 1187 | |||

Brd7
Genomic Map