siRNA Details
| Gene Symbol: Phkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014152.1 | NP_001014174.1 | 13 | 1264 | ||
| XM_006255269.2 | XP_006255331.1 | 13 | 1301 | |||
| XM_006255270.2 | XP_006255332.1 | 13 | 1301 | |||
| XM_008772410.1 | XP_008770632.1 | 13 | 2013 | |||
| GenBank mRNA | BC085909.1 | - | 13 | 1264 | 7096506 | |

Phkb
Genomic Map