siRNA Details
| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772368.1 | XP_008770590.1 | 1 | 64 | ||
| XM_008772369.1 | XP_008770591.1 | 1 | 64 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asna1| Gene Symbol: Asna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772368.1 | XP_008770590.1 | 1 | 64 | ||
| XM_008772369.1 | XP_008770591.1 | 1 | 64 | |||