siRNA Details
| Gene Symbol: Rad23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013190.1 | NP_001013208.1 | 5, 6 | 653 | ||
| XM_006255276.2 | XP_006255338.1 | 5, 6 | 714 | |||
| XM_006255277.2 | XP_006255339.1 | 5, 6 | 714 | |||
| XM_008772411.1 | XP_008770633.1 | 5, 6 | 1969 | |||
| GenBank mRNA | BC084695.1 | - | 5, 6 | 653 | 7114647 | |

Rad23a
Genomic Map