siRNA Details
| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108441.1 | NP_001101911.1 | 4 | 1004 | ||
| XM_006255282.2 | XP_006255344.1 | 3 | 1038 | |||
| GenBank mRNA | BC159430.1 | - | 3 | 934 | 9089687 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb1| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108441.1 | NP_001101911.1 | 4 | 1004 | ||
| XM_006255282.2 | XP_006255344.1 | 3 | 1038 | |||
| GenBank mRNA | BC159430.1 | - | 3 | 934 | 9089687 | |