siRNA Details
| Gene Symbol: Ranbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135875.1 | NP_001129347.1 | 13 | 1881 | ||
| XM_008772517.1 | XP_008770739.1 | 12 | 1896 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ranbp10| Gene Symbol: Ranbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135875.1 | NP_001129347.1 | 13 | 1881 | ||
| XM_008772517.1 | XP_008770739.1 | 12 | 1896 | |||