siRNA Details
| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108453.1 | NP_001101923.1 | 10 | 1458 | ||
| XM_006255770.2 | XP_006255832.1 | 9 | 2352 | |||
| XM_006255772.2 | XP_006255834.1 | 10 | 1613 | |||
| XM_008772629.1 | XP_008770851.1 | 9 | 3323 | |||
| XM_008772630.1 | XP_008770852.1 | 8 | 3246 | |||
| XM_008772631.1 | XP_008770853.1 | 9 | 1808 | |||

Cbfa2t3
Genomic Map