siRNA Details
| Gene Symbol: Cpne7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108454.1 | NP_001101924.1 | 11 | 1333 | ||
| XM_006255774.2 | XP_006255836.1 | 14 | 1596 | |||
| XM_008772632.1 | XP_008770854.1 | 14 | 1980 | |||
| GenBank mRNA | AM747283.1 | - | 14 | 1428 | ||

Cpne7
Genomic Map