siRNA Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108455.1 | NP_001101925.1 | 35 | 3522 | ||
| XM_008772633.1 | XP_008770855.1 | 36 | 3646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fanca| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108455.1 | NP_001101925.1 | 35 | 3522 | ||
| XM_008772633.1 | XP_008770855.1 | 36 | 3646 | |||