siRNA Details
| Gene Symbol: Ltv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014157.1 | NP_001014179.1 | 10, 11 | 1321 | ||
| GenBank mRNA | BC079397.1 | - | 10, 11 | 1321 | 7112501 | |
| FQ213117.1 | - | 5, 6 | 757 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltv1| Gene Symbol: Ltv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014157.1 | NP_001014179.1 | 10, 11 | 1321 | ||
| GenBank mRNA | BC079397.1 | - | 10, 11 | 1321 | 7112501 | |
| FQ213117.1 | - | 5, 6 | 757 | |||