siRNA Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100735.1 | NP_001094205.1 | 14 | 1702 | ||
| GenBank mRNA | AB244983.2 | - | 13 | 1430 | ||
| CK365681.1 | - | 1 | 124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpcat1| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100735.1 | NP_001094205.1 | 14 | 1702 | ||
| GenBank mRNA | AB244983.2 | - | 13 | 1430 | ||
| CK365681.1 | - | 1 | 124 | |||