siRNA Details
Gene Symbol: Eps8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108467.1 | NP_001101937.1 | 10 | 1173 | ||
XM_006228278.2 | XP_006228340.1 | 11 | 1452 | |||
XM_006228279.2 | XP_006228341.1 | 11 | 1349 | |||
XM_006228280.2 | XP_006228342.1 | 11 | 1224 | |||
XM_006228282.2 | XP_006228344.1 | 11 | 1348 | |||
XM_008758941.1 | XP_008757163.1 | 11 | 1245 | |||
XM_008758942.1 | XP_008757164.1 | 11 | 1102 | |||
XM_008758943.1 | XP_008757165.1 | 11 | 1452 |
