siRNA Details
Gene Symbol: Eps8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108467.1 | NP_001101937.1 | 15 | 1715 | ||
XM_006228278.2 | XP_006228340.1 | 16 | 1994 | |||
XM_006228279.2 | XP_006228341.1 | 16 | 1891 | |||
XM_006228280.2 | XP_006228342.1 | 16 | 1766 | |||
XM_006228282.2 | XP_006228344.1 | 16 | 1890 | |||
XM_008758941.1 | XP_008757163.1 | 16 | 1787 | |||
XM_008758942.1 | XP_008757164.1 | 16 | 1644 | |||
XM_008758943.1 | XP_008757165.1 | 16 | 1994 |
