siRNA Details
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037790.2 | NP_001032879.2 | 4 | 435 | ||
XM_006228218.1 | XP_006228280.1 | 4 | 609 | |||
XM_006228219.2 | XP_006228281.1 | 4 | 766 | |||
XM_006228220.1 | XP_006228282.1 | 4 | 609 | |||
XM_008758946.1 | XP_008757168.1 | 4 | 609 | |||
XM_008758947.1 | XP_008757169.1 | 4 | 609 | |||
XM_008758948.1 | XP_008757170.1 | 4 | 609 |
