siRNA Details
| Gene Symbol: Hnrnpul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108477.1 | NP_001101947.1 | 1, 2 | 385 | ||
| XM_008758955.1 | XP_008757177.1 | 1, 2 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hnrnpul1| Gene Symbol: Hnrnpul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108477.1 | NP_001101947.1 | 1, 2 | 385 | ||
| XM_008758955.1 | XP_008757177.1 | 1, 2 | 377 | |||