siRNA Details
| Gene Symbol: Paf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024898.1 | NP_001020069.1 | 13 | 1358 | ||
| GenBank mRNA | BC097494.1 | - | 13 | 1358 | 7461781 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Paf1| Gene Symbol: Paf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024898.1 | NP_001020069.1 | 13 | 1358 | ||
| GenBank mRNA | BC097494.1 | - | 13 | 1358 | 7461781 | |