siRNA Details
| Gene Symbol: Prodh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038588.1 | NP_001033677.1 | 2 | 248 | ||
| GenBank mRNA | AF222852.1 | - | 2 | 238 | ||
| BC158806.1 | - | 2 | 248 | 7324442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prodh2| Gene Symbol: Prodh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038588.1 | NP_001033677.1 | 2 | 248 | ||
| GenBank mRNA | AF222852.1 | - | 2 | 238 | ||
| BC158806.1 | - | 2 | 248 | 7324442 | ||