siRNA Details
| Gene Symbol: Haus5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728731.3 | XP_002728777.1 | 15 | 1427 | ||
| XM_008759256.1 | XP_008757478.1 | 15 | 1419 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Haus5| Gene Symbol: Haus5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728731.3 | XP_002728777.1 | 15 | 1427 | ||
| XM_008759256.1 | XP_008757478.1 | 15 | 1419 | |||