siRNA Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127552.1 | NP_001121024.1 | 2, 3 | 386 | ||
| XM_006228856.2 | XP_006228918.1 | 2, 3 | 434 | |||
| XM_006228857.2 | XP_006228919.1 | 2, 3 | 434 | |||
| XM_006228858.2 | XP_006228920.1 | 2, 3 | 434 | |||
| GenBank mRNA | BC166537.1 | - | 2, 3 | 386 | 7191211 | |

Lsm14a
Genomic Map