siRNA Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005561.1 | NP_001005561.1 | 14 | 1409 | ||
| XM_006229095.2 | XP_006229157.1 | 14 | 1468 | |||
| GenBank mRNA | BC081928.1 | - | 14 | 1409 | 7113277 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vrk3| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005561.1 | NP_001005561.1 | 14 | 1409 | ||
| XM_006229095.2 | XP_006229157.1 | 14 | 1468 | |||
| GenBank mRNA | BC081928.1 | - | 14 | 1409 | 7113277 | |