siRNA Details
| Gene Symbol: Kdelr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017385.1 | NP_001017385.1 | 3 | 381 | ||
| GenBank mRNA | BC092600.1 | - | 3 | 381 | 7121304 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdelr1| Gene Symbol: Kdelr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017385.1 | NP_001017385.1 | 3 | 381 | ||
| GenBank mRNA | BC092600.1 | - | 3 | 381 | 7121304 | |