siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014161.1 | NP_001014183.1 | 5 | 650 | ||
| GenBank mRNA | BC076398.1 | - | 5 | 650 | 7110062 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Idh2| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014161.1 | NP_001014183.1 | 5 | 650 | ||
| GenBank mRNA | BC076398.1 | - | 5 | 650 | 7110062 | |